missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-nitrobenzoic acid, 98+%

Catalog No. p-7044963
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0889106 5 g
AAL0889114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0889106 Supplier Thermo Scientific Chemicals Supplier No. L0889106

CAS: 5081-36-7 | C8H7NO5 | 197.146 g/mol

3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5081-36-7
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007353
InChI Key PWURRRRGLCVBMX-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid, 4-nitro-3-methoxybenzoic acid, 4-nitro-m-anisic acid, benzoic acid, 3-methoxy-4-nitro, 3-methoxy-4-nitrobenzoicacid, pubchem3918, acmc-1bn9s, ksc270m6t, 3-methoxy4-nitro-benzoic acid, 3-methoxy-4-nitro-benzoic acid
PubChem CID 78764
SMILES COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 231°C to 235°C
Quantity 5 g
Beilstein 2696042
Solubility Information Insoluble in water.
Formula Weight 197.15
Percent Purity ≥98%
Chemical Name or Material 3-Methoxy-4-nitrobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 225-793-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only