missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-methylaniline, 99+%

Catalog No. p-7033278
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50362MD 250 mg
AAH5036203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50362MD Supplier Thermo Scientific Chemicals Supplier No. H50362MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16452-01-0 | C8H11NO | 137.18 g/mol

3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 16452-01-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00025371
InChI Key ONADZNBSLRAJFW-UHFFFAOYSA-N
Synonym o-cresidine, benzenamine, 3-methoxy-4-methyl, 3-methoxy-4-methyl aniline, ccris 4606, unii-v82scc5x3q, 5-amino-2-methylanisole, 5-amino-2-methyl-anisol, 2-methoxy-4-aminotoluene, 3-methoxy-4-methyl-aniline, v82scc5x3q
PubChem CID 27882
IUPAC Name 3-methoxy-4-methylaniline
SMILES COC1=CC(N)=CC=C1C

Specifications

Melting Point 56°C to 60°C
Quantity 250 mg
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 137.18
Percent Purity ≥99%
Chemical Name or Material 3-Methoxy-4-methylaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 240-500-8
RTECSNumber CY0888000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only