CAS RN 16452-01-0
IUPAC Name:
3-methoxy-4-methylaniline
Synonyms:
5-amino-2-methyl-anisol
Molecular Weight (g/mol):
137.18
Molecular Formula:
C8H11NO
InChi Key:
ONADZNBSLRAJFW-UHFFFAOYSA-N
SMILES:
COC1=CC(N)=CC=C1C
1
–
1
of
1
results
1
Promotions Available
3-Methoxy-4-methylaniline, 99+%
3-Methoxy-4-methylaniline, 99+%
CAS: 16452-01-0 Molecular Formula: C8H11NO Molecular Weight (g/mol): 137.18 MDL Number: MFCD00025371 InChI Key: ONADZNBSLRAJFW-UHFFFAOYSA-N Synonym: o-cresidine,benzenamine, 3-methoxy-4-methyl,3-methoxy-4-methyl aniline,ccris 4606,unii-v82scc5x3q,5-amino-2-methylanisole,5-amino-2-methyl-anisol,2-methoxy-4-aminotoluene,3-methoxy-4-methyl-aniline,v82scc5x3q PubChem CID: 27882 IUPAC Name: 3-methoxy-4-methylaniline SMILES: COC1=CC(N)=CC=C1C