missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-nitrobenzoic Acid 99.0+%, TCI America™

$83.89 - $460.43

Chemical Identifiers

CAS 4920-80-3
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007143
InChI Key YMOMYSDAOXOCID-UHFFFAOYSA-N
Synonym 3-methoxy-2-nitrobenzoic acid, 2-nitro-3-methoxybenzoic acid, benzoic acid, 3-methoxy-2-nitro, 2-nitro-m-anisic acid, m-anisic acid, 2-nitro, 3-methoxy-2-nitrobenzoicacid, pubchem12905, acmc-1ao65, ksc238g2p
PubChem CID 78632
SMILES COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10361G
View Documents
TCI America
M10361G
1 g
Each for $83.89
Only null left
 
M103610G
View Documents
TCI America
M103610G
10 g
Each for $460.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4920-80-3
197.146
YMOMYSDAOXOCID-UHFFFAOYSA-N
78632
C8H7NO5
MFCD00007143
3-methoxy-2-nitrobenzoic acid, 2-nitro-3-methoxybenzoic acid, benzoic acid, 3-methoxy-2-nitro, 2-nitro-m-anisic acid, m-anisic acid, 2-nitro, 3-methoxy-2-nitrobenzoicacid, pubchem12905, acmc-1ao65, ksc238g2p
COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O

Specifications

4920-80-3
C8H7NO5
1 g
YMOMYSDAOXOCID-UHFFFAOYSA-N
197.146
197.15
Crystalline Powder
White
MFCD00007143
3-methoxy-2-nitrobenzoic acid, 2-nitro-3-methoxybenzoic acid, benzoic acid, 3-methoxy-2-nitro, 2-nitro-m-anisic acid, m-anisic acid, 2-nitro, 3-methoxy-2-nitrobenzoicacid, pubchem12905, acmc-1ao65, ksc238g2p
COC1=CC=CC(=C1[N+](=O)[O-])C(=O)O
78632
≥99.0% (T)
3-Methoxy-2-nitrobenzoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'