accessibility menu, dialog, popup

UserName

CAS RN 4920-80-3

CH3OOHONOO

CAS RN 4920-80-3

IUPAC Name: 3-methoxy-2-nitrobenzoic acid
Synonyms: 3-methoxy-2-nitrobenzoic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: YMOMYSDAOXOCID-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results