missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-2-methylbenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 149°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 166.18 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Methoxy-2-methylbenzoic Acid |
Safety and Handling
| TSCA | No |