missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-1-butanol 98.0+%, TCI America™
$37.29 - $57.79
Chemical Identifiers
| CAS | 2517-43-3 |
|---|---|
| Molecular Formula | C5H12O2 |
| Molecular Weight (g/mol) | 104.15 |
| MDL Number | MFCD00002931 |
| InChI Key | JSGVZVOGOQILFM-UHFFFAOYNA-N |
| Synonym | 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol |
| PubChem CID | 17291 |
| IUPAC Name | 3-methoxybutan-1-ol |
| SMILES | COC(C)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M010925ML
|
TCI America
M010925ML |
25 mL |
Each for $37.29
|
|
|||||
|
M0109500ML
|
TCI America
M0109500ML |
500 mL |
Each for $57.79
|
|
|||||
Chemical Identifiers
| 2517-43-3 | |
| 104.15 | |
| JSGVZVOGOQILFM-UHFFFAOYNA-N | |
| 17291 | |
| COC(C)CCO |
| C5H12O2 | |
| MFCD00002931 | |
| 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol | |
| 3-methoxybutan-1-ol |
Specifications
| 2517-43-3 | |
| 158°C | |
| MFCD00002931 | |
| 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol | |
| COC(C)CCO | |
| 104.15 | |
| 104.15 | |
| Liquid |
| Colorless | |
| C5H12O2 | |
| 25 mL | |
| JSGVZVOGOQILFM-UHFFFAOYNA-N | |
| 3-methoxybutan-1-ol | |
| 17291 | |
| ≥98.0% (GC) | |
| 3-Methoxy-1-butanol |
Safety and Handling
EINECSNumber : (2)-0409
TSCA : Yes