missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxy-1-butanol 98.0+%, TCI America™
Supplier: TCI America M010925ML
Specifications
| 3-Methoxy-1-butanol | |
| Colorless | |
| C5H12O2 | |
| 25 mL | |
| JSGVZVOGOQILFM-UHFFFAOYNA-N | |
| 3-methoxybutan-1-ol | |
| 17291 | |
| ≥98.0% (GC) |
| 2517-43-3 | |
| 158°C | |
| MFCD00002931 | |
| 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol | |
| COC(C)CCO | |
| 104.15 | |
| 104.15 | |
| Liquid |
Chemical Identifiers
| 2517-43-3 | |
| 104.15 | |
| JSGVZVOGOQILFM-UHFFFAOYNA-N | |
| 17291 | |
| COC(C)CCO |
| C5H12O2 | |
| MFCD00002931 | |
| 3-methoxy-1-butanol, 3-methoxybutanol, 1-butanol, 3-methoxy, methoxybutanol, ccris 8976, 1-butanol, 3-methoxy-,-, 1-butanol, 3-methoxy-, r, 3-methoxy-butanol, 3-methoxybutane-1-ol, 3-methoxybutyl alcohol | |
| 3-methoxybutan-1-ol |
Safety and Handling
EINECSNumber : (2)-0409
TSCA : Yes