accessibility menu, dialog, popup

UserName

H3COCH3OH

CAS RN 2517-43-3

IUPAC Name: 3-methoxybutan-1-ol
Synonyms: 3-methoxybutanol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: JSGVZVOGOQILFM-UHFFFAOYNA-N
SMILES: COC(C)CCO
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)