missing translation for 'onlineSavingsMsg'
Learn More

3-Isocyanato-5-methyl-2-phenylfuran, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC49706DA

 View more versions of this product

Catalog No. CC49706DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Isocyanato-5-methyl-2-phenylfuran
96.5
97%
C12H9NO2
1g
GDLFJANXKGIZKA-UHFFFAOYSA-N
3-isocyanato-5-methyl-2-phenylfuran
18525836
97%
898289-18-4
100.0
Amber Glass Bottle
MFCD09025851
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
199.209
199.21

Chemical Identifiers

898289-18-4
199.209
GDLFJANXKGIZKA-UHFFFAOYSA-N
18525836
CC1=CC(=C(O1)C2=CC=CC=C2)N=C=O
C12H9NO2
MFCD09025851
5-methyl-2-phenylfuran-3-isocyanate, furan,3-isocyanato-5-methyl-2-phenyl
3-isocyanato-5-methyl-2-phenylfuran
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions