missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodopropionic Acid 98.0+%, TCI America™
Supplier: TCI America I00715G
Specifications
| 3-Iodopropionic Acid | |
| 82°C | |
| MFCD00002765 | |
| 3261 | |
| KMRNTNDWADEIIX-UHFFFAOYSA-N | |
| 3-iodopropanoic acid | |
| 8856 | |
| ≥98.0% (GC,T) |
| 141-76-4 | |
| C3H5IO2 | |
| 5 g | |
| 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid | |
| OC(=O)CCI | |
| 199.98 | |
| 199.98 | |
| Crystalline Powder |
Chemical Identifiers
| 141-76-4 | |
| 199.98 | |
| KMRNTNDWADEIIX-UHFFFAOYSA-N | |
| 8856 | |
| OC(=O)CCI |
| C3H5IO2 | |
| MFCD00002765 | |
| 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid | |
| 3-iodopropanoic acid |
Safety and Handling
RTECSNumber : UF4900000
TSCA : Yes