missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopropionic Acid 98.0+%, TCI America™

Catalog No. p-7131195
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00715G 5 g
I007125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00715G Supplier TCI America Supplier No. I00715G
Only null left

Chemical Identifiers

CAS 141-76-4
Molecular Formula C3H5IO2
Molecular Weight (g/mol) 199.98
MDL Number MFCD00002765
InChI Key KMRNTNDWADEIIX-UHFFFAOYSA-N
Synonym 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid
PubChem CID 8856
IUPAC Name 3-iodopropanoic acid
SMILES OC(=O)CCI

Specifications

Melting Point 82°C
Quantity 5 g
UN Number 3261
Formula Weight 199.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodopropionic Acid
RTECSNumber UF4900000
TSCA Yes