accessibility menu, dialog, popup

UserName

OHOI

CAS RN 141-76-4

IUPAC Name: 3-iodopropanoic acid
Synonyms: 3-iodopropionic acid
Molecular Weight (g/mol): 199.98
Molecular Formula: C3H5IO2
InChi Key: KMRNTNDWADEIIX-UHFFFAOYSA-N
SMILES: OC(=O)CCI
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)