Learn More
3-Iodophenol, 98%
CAS: 626-02-8 | C6H5IO | 220.009 g/mol
Supplier: Thermo Scientific Chemicals A1115518
Description
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.
Solubility
Slightly soluble in water.
Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 3-Iodophenol | |
| 38°C to 43°C | |
| C6H5IO | |
| 50 g | |
| Light sensitive | |
| Slightly soluble in water. | |
| C1=CC(=CC(=C1)I)O | |
| 220.009 | |
| CHEBI:33439 | |
| 98% |
| 626-02-8 | |
| >110°C (230°F) | |
| MFCD00002261 | |
| 2039304 | |
| m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968 | |
| FXTKWBZFNQHAAO-UHFFFAOYSA-N | |
| 3-iodophenol | |
| 12272 | |
| 220.01 |
Chemical Identifiers
| 626-02-8 | |
| 220.009 | |
| FXTKWBZFNQHAAO-UHFFFAOYSA-N | |
| 12272 | |
| 3-iodophenol |
| C6H5IO | |
| MFCD00002261 | |
| m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968 | |
| CHEBI:33439 | |
| C1=CC(=CC(=C1)I)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-923-2
RTECSNumber : SL5490000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only