missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol, 98%

Catalog No. p-7081253
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1115509 10 g
AAA1115518 50 g
AAA1115530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1115509 Supplier Thermo Scientific Chemicals Supplier No. A1115509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-02-8 | C6H5IO | 220.009 g/mol

3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenol is used in esterification, electrophilic aromatic substitution, alkylation, nucleophilic aromatic substitution, Mitsunobu, transition metal-catalyzed cross-coupling, acylation, and oxidation reactions.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 626-02-8
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.009
MDL Number MFCD00002261
InChI Key FXTKWBZFNQHAAO-UHFFFAOYSA-N
Synonym m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
PubChem CID 12272
ChEBI CHEBI:33439
IUPAC Name 3-iodophenol
SMILES C1=CC(=CC(=C1)I)O

Specifications

Melting Point 38°C to 43°C
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 2039304
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 3-Iodophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-923-2
RTECSNumber SL5490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only