missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzylamine Hydrochloride 98.0+%, TCI America™

Catalog No. I07101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07101G 1 g
I07105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07101G Supplier TCI America Supplier No. I07101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3718-88-5
Molecular Formula C7H9ClIN
Molecular Weight (g/mol) 269.51
MDL Number MFCD00012857
InChI Key PYFDZOCGFHIRST-UHFFFAOYSA-N
Synonym 3-iodobenzylamine hydrochloride, 3-iodophenyl methanamine hydrochloride, 3-iodobenzylamine hcl, 3-iodobenzylaminehydrochloride, benzenemethanamine, 3-iodo-, hydrochloride, 1-3-iodophenyl methanamine hydrochloride, pubchem3302, acmc-1cmc7, 3-iodobenzylamine hcl salt, ksc491e3h
PubChem CID 2723861
IUPAC Name (3-iodophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)I)CN.Cl

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 1 g
Formula Weight 269.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzylamine Hydrochloride