accessibility menu, dialog, popup

UserName

HClH2NI

CAS RN 3718-88-5

IUPAC Name: hydrogen 1-(3-iodophenyl)methanamine chloride
Synonyms: hydrogen 1-(3-iodophenyl)methanamine chloride
Molecular Weight (g/mol): 269.51
Molecular Formula: C7H9ClIN
InChi Key: PYFDZOCGFHIRST-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=CC(I)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results