missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzyl Cyanide 96.0+%, TCI America™

Catalog No. I10021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10021G 1 g
I10025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10021G Supplier TCI America Supplier No. I10021G

Chemical Identifiers

CAS 130723-54-5
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.05
MDL Number MFCD00040890
InChI Key LVOKGAHCTHNWIL-UHFFFAOYSA-N
Synonym 3-iodophenylacetonitrile, 2-3-iodophenyl acetonitrile, 3-iodobenzyl cyanide, 3-iodophenyl acetonitrile, m-iodobenzyl cyanide, 3-iodophenyacetonitrile, pubchem12771, 3-iodo-1-phenylacetonitrile, acmc-1c0v0, benzeneacetonitrile, 3-iodo
PubChem CID 2759368
IUPAC Name 2-(3-iodophenyl)acetonitrile
SMILES IC1=CC=CC(CC#N)=C1

Specifications

Melting Point 36°C
Boiling Point 138°C
Quantity 1 g
UN Number 3439
Formula Weight 243.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Iodobenzyl Cyanide
TSCA No
Recommended Storage Refrigerator