missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodobenzyl Bromide 97.0+%, TCI America™
$144.39 - $564.08
Chemical Identifiers
| CAS | 49617-83-6 |
|---|---|
| Molecular Formula | C7H6BrI |
| Molecular Weight (g/mol) | 296.933 |
| InChI Key | BACZSVQZBSCWIG-UHFFFAOYSA-N |
| Synonym | 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide |
| PubChem CID | 2759361 |
| IUPAC Name | 1-(bromomethyl)-3-iodobenzene |
| SMILES | C1=CC(=CC(=C1)I)CBr |
Chemical Identifiers
| 49617-83-6 | |
| 296.933 | |
| 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide | |
| 1-(bromomethyl)-3-iodobenzene |
| C7H6BrI | |
| BACZSVQZBSCWIG-UHFFFAOYSA-N | |
| 2759361 | |
| C1=CC(=CC(=C1)I)CBr |
Specifications
| 49617-83-6 | |
| C7H6BrI | |
| 3261 | |
| BACZSVQZBSCWIG-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-iodobenzene | |
| 2759361 | |
| ≥97.0% (GC) | |
| 3-Iodobenzyl Bromide |
| 52°C | |
| 5 g | |
| 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide | |
| C1=CC(=CC(=C1)I)CBr | |
| 296.933 | |
| 296.93 | |
| Crystalline Powder |
Safety and Handling
TSCA : No