accessibility menu, dialog, popup

UserName

IBr

CAS RN 49617-83-6

IUPAC Name: 1-(bromomethyl)-3-iodobenzene
Synonyms: 1-(bromomethyl)-3-iodobenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: BACZSVQZBSCWIG-UHFFFAOYSA-N
SMILES: BrCC1=CC(I)=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)