missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzyl bromide, 97%

Catalog No. AC392530250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392530250 25 g Glass bottle
AC392530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392530250 Supplier Thermo Scientific Chemicals Supplier No. 392530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 49617-83-6 | C7H6BrI | 296.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49617-83-6
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD00019016
InChI Key BACZSVQZBSCWIG-UHFFFAOYSA-N
Synonym 3-iodobenzyl bromide, 1-bromomethyl-3-iodobenzene, alpha-bromo-3-iodotoluene, 3-iodobenzylbromide, m-iodobenzyl bromide, benzene, 1-bromomethyl-3-iodo, m-iodobenzylbromide, 3-iodo-benzylbromide, 3-iodo-benzyl bromide, meta-iodobenzyl bromide
PubChem CID 2759361
IUPAC Name 1-(bromomethyl)-3-iodobenzene
SMILES C1=CC(=CC(=C1)I)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 50°C
Color Brown or Orange to Red
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4I
Quantity 25 g
Formula Weight 296.93
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only