missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodobenzonitrile 98.0+%, TCI America™
$87.60 - $260.97
Chemical Identifiers
| CAS | 69113-59-3 |
|---|---|
| Molecular Formula | C7H4IN |
| Molecular Weight (g/mol) | 229.02 |
| MDL Number | MFCD00079762 |
| InChI Key | BGARPMGQRREXLN-UHFFFAOYSA-N |
| Synonym | m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930 |
| PubChem CID | 144341 |
| IUPAC Name | 3-iodobenzonitrile |
| SMILES | C1=CC(=CC(=C1)I)C#N |
Chemical Identifiers
| 69113-59-3 | |
| 229.02 | |
| BGARPMGQRREXLN-UHFFFAOYSA-N | |
| 144341 | |
| C1=CC(=CC(=C1)I)C#N |
| C7H4IN | |
| MFCD00079762 | |
| m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930 | |
| 3-iodobenzonitrile |
Specifications
| 69113-59-3 | |
| C7H4IN | |
| 5 g | |
| m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930 | |
| C1=CC(=CC(=C1)I)C#N | |
| 229.02 | |
| 229.02 | |
| Crystalline Powder |
| 41°C | |
| MFCD00079762 | |
| 3439 | |
| BGARPMGQRREXLN-UHFFFAOYSA-N | |
| 3-iodobenzonitrile | |
| 144341 | |
| ≥98.0% (GC) | |
| 3-Iodobenzonitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator