missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Iodobenzonitrile 98.0+%, TCI America™
Supplier: TCI America I066025G
Specifications
3-Iodobenzonitrile | |
41°C | |
MFCD00079762 | |
3439 | |
BGARPMGQRREXLN-UHFFFAOYSA-N | |
3-iodobenzonitrile | |
144341 | |
≥98.0% (GC) |
69113-59-3 | |
C7H4IN | |
25 g | |
m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930 | |
C1=CC(=CC(=C1)I)C#N | |
229.02 | |
229.02 | |
Crystalline Powder |
Chemical Identifiers
69113-59-3 | |
229.02 | |
BGARPMGQRREXLN-UHFFFAOYSA-N | |
144341 | |
C1=CC(=CC(=C1)I)C#N |
C7H4IN | |
MFCD00079762 | |
m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930 | |
3-iodobenzonitrile |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator