missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoaniline, 98%

CAS: 626-01-7 | C6H6IN | 219.025 g/mol

$117.40 - $409.55

Chemical Identifiers

CAS 626-01-7
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007781
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
PubChem CID 12271
IUPAC Name 3-iodoaniline
SMILES C1=CC(=CC(=C1)I)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0115909
View Documents
Thermo Scientific Chemicals
L0115909
10 g
Each for $117.40
Only null left
 
AAL0115918
View Documents
Thermo Scientific Chemicals
L0115918
50 g
Each for $409.55
Only null left
 
Description

Description

3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
Specifications

Chemical Identifiers

626-01-7
219.025
FFCSRWGYGMRBGD-UHFFFAOYSA-N
12271
C1=CC(=CC(=C1)I)N
C6H6IN
MFCD00007781
m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
3-iodoaniline

Specifications

626-01-7
1.821
>110°C (230°F)
1.684
10 g
Light sensitive
Insoluble in water.
C1=CC(=CC(=C1)I)N
219.025
219.03
3-Iodoaniline
21°C to 24°C
145°C to 146°C (15 mmHg)
C6H6IN
MFCD00007781
636058
m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
FFCSRWGYGMRBGD-UHFFFAOYSA-N
3-iodoaniline
12271
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-922-7

RTECSNumber : BY3750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only