missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Iodo-L-tyrosine, 97%

CAS: 70-78-0 | C9H10INO3 | 307.09 g/mol

$314.14

Chemical Identifiers

CAS 70-78-0
Molecular Formula C9H10INO3
Molecular Weight (g/mol) 307.09
MDL Number MFCD00002608
InChI Key UQTZMGFTRHFAAM-JLDDOWRYNA-N
Synonym 3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
PubChem CID 439744
ChEBI CHEBI:27847
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC122440050
View Documents Promotion Details
Thermo Scientific Chemicals
122440050
5 g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70-78-0
307.09
UQTZMGFTRHFAAM-JLDDOWRYNA-N
439744
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
C9H10INO3
MFCD00002608
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
CHEBI:27847
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O

Specifications

196.0°C to 198.0°C
96.0
White
Authentic
C9H10INO3
− 3.18
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
1% max. (K.F.)
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
307.09
CHEBI:27847
97%
3-Iodo-L-tyrosine
70-78-0
100.0
20ppm max.
97%
MFCD00002608
15, 5090
Solubility in water: .. Other solubilities: soluble in 15 parts boiling water
UQTZMGFTRHFAAM-JLDDOWRYNA-N
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
439744
307.08
Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only