accessibility menu, dialog, popup

UserName

(S)ONH2OHHOI

CAS RN 70-78-0

IUPAC Name: (2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
Synonyms: 3-iodo-tyrosine
Molecular Weight (g/mol): 307.09
Molecular Formula: C9H10INO3
InChi Key: UQTZMGFTRHFAAM-JLDDOWRYNA-N
SMILES: N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (4)
  • (1)
  • (4)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)