missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methoxybenzoic Acid 97.0+%, TCI America™

Catalog No. I09595G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I09595G Supplier TCI America Supplier No. I09595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68507-19-7
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00017347
InChI Key HNJSYSWRPCCSQI-UHFFFAOYSA-N
Synonym 3-iodo-4-methoxy-benzoic acid, 3-iodo-p-anisic acid, benzoic acid, 3-iodo-4-methoxy, 3-iodo-4-methoxybenzoicacid, pubchem4973, acmc-209o3r, akos bbb/204, benzoic acid,3-iodo-4-methoxy, 3-iodo-4-methoxybenzoic acid
PubChem CID 624158
IUPAC Name 3-iodo-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)I

Specifications

Melting Point 243°C
Quantity 5 g
Formula Weight 278.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-4-methoxybenzoic Acid
TSCA No
Recommended Storage Refrigerator