accessibility menu, dialog, popup

UserName

H3COOHOI

CAS RN 68507-19-7

IUPAC Name: 3-iodo-4-methoxybenzoic acid
Synonyms: 3-iodo-4-methoxybenzoic acid
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: HNJSYSWRPCCSQI-UHFFFAOYSA-N
SMILES: COC1=C(I)C=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results