missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Indolepropionic Acid 98.0+%, TCI America™
Supplier: TCI America I00325G
Specifications
| 3-Indolepropionic Acid | |
| 132°C | |
| MFCD00005660 | |
| 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid | |
| OC(=O)CCC1=CNC2=CC=CC=C12 | |
| 189.21 | |
| CHEBI:43580 | |
| ≥98.0% (T) |
| 830-96-6 | |
| C11H11NO2 | |
| 5 g | |
| GOLXRNDWAUTYKT-UHFFFAOYSA-N | |
| 3-(1H-indol-3-yl)propanoic acid | |
| 3744 | |
| 189.21 | |
| Crystalline Powder |
Chemical Identifiers
| 830-96-6 | |
| 189.21 | |
| GOLXRNDWAUTYKT-UHFFFAOYSA-N | |
| 3744 | |
| 3-(1H-indol-3-yl)propanoic acid |
| C11H11NO2 | |
| MFCD00005660 | |
| 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid | |
| CHEBI:43580 | |
| OC(=O)CCC1=CNC2=CC=CC=C12 |
Safety and Handling
RTECSNumber : NM1329000
TSCA : Yes