missing translation for 'onlineSavingsMsg'
Learn More

3-Indolepropionic Acid 98.0+%, TCI America™

Catalog No. p-7131170
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I00325G 5 g
I003225G 25 g
2 options

Catalog No. I00325G

Supplier: TCI America I00325G

Only null left

Chemical Identifiers

CAS 830-96-6
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005660
InChI Key GOLXRNDWAUTYKT-UHFFFAOYSA-N
Synonym 3-indolepropionic acid, indole-3-propionic acid, 3-1h-indol-3-yl propanoic acid, 1h-indole-3-propanoic acid, indolepropionic acid, indolylpropionic acid, 3-3-indolyl propionic acid, 3-3-indolyl propanoic acid, ipa auxin, 1h-indole-3-propionic acid
PubChem CID 3744
ChEBI CHEBI:43580
IUPAC Name 3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)CCC1=CNC2=CC=CC=C12

Specifications

Melting Point 132°C
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indolepropionic Acid
RTECSNumber NM1329000
TSCA Yes