missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine-2-carboxylic acid, 98%

CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol

$88.99 - $225.96

Chemical Identifiers

CAS 874-24-8
Molecular Formula C6H4NO3
Molecular Weight (g/mol) 138.10
MDL Number MFCD00006294
InChI Key BRARRAHGNDUELT-UHFFFAOYSA-M
Synonym 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
PubChem CID 13401
ChEBI CHEBI:64342
IUPAC Name 3-hydroxypyridine-2-carboxylic acid
SMILES OC1=CC=CN=C1C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0748903
View Documents
Thermo Scientific Chemicals
L0748903
1 g
Each for $88.99
Only null left
 
AAL0748906
View Documents
Thermo Scientific Chemicals
L0748906
5 g
Each for $225.96
Only null left
 
Description

Description

3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.

Solubility
Soluble in water (0.25g in 10ml).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

874-24-8
138.10
BRARRAHGNDUELT-UHFFFAOYSA-M
13401
3-hydroxypyridine-2-carboxylic acid
C6H4NO3
MFCD00006294
3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
CHEBI:64342
OC1=CC=CN=C1C([O-])=O

Specifications

874-24-8
C6H4NO3
1 g
3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
BRARRAHGNDUELT-UHFFFAOYSA-M
3-hydroxypyridine-2-carboxylic acid
13401
139.11
3-Hydroxypyridine-2-carboxylic acid
∽214°C (decomposition)
MFCD00006294
118954
Soluble in water (0.25g in 10ml).
OC1=CC=CN=C1C([O-])=O
138.10
CHEBI:64342
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

EINECSNumber : 212-859-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only