Learn More
3-Hydroxypyridine-2-carboxylic acid, 98%
CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol
Supplier: Thermo Scientific Chemicals L0748906
Description
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.
Solubility
Soluble in water (0.25g in 10ml).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| 3-Hydroxypyridine-2-carboxylic acid | |
| ∽214°C (decomposition) | |
| MFCD00006294 | |
| 118954 | |
| Soluble in water (0.25g in 10ml). | |
| OC1=CC=CN=C1C([O-])=O | |
| 138.10 | |
| CHEBI:64342 | |
| 98% |
| 874-24-8 | |
| C6H4NO3 | |
| 5 g | |
| 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid | |
| BRARRAHGNDUELT-UHFFFAOYSA-M | |
| 3-hydroxypyridine-2-carboxylic acid | |
| 13401 | |
| 139.11 |
Chemical Identifiers
| 874-24-8 | |
| 138.10 | |
| BRARRAHGNDUELT-UHFFFAOYSA-M | |
| 13401 | |
| 3-hydroxypyridine-2-carboxylic acid |
| C6H4NO3 | |
| MFCD00006294 | |
| 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid | |
| CHEBI:64342 | |
| OC1=CC=CN=C1C([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 212-859-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only