accessibility menu, dialog, popup

UserName

OHNOO

CAS RN 874-24-8

IUPAC Name: 3-hydroxypyridine-2-carboxylate
Synonyms: 3-hydroxypicolinic acid
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H4NO3
InChi Key: BRARRAHGNDUELT-UHFFFAOYSA-M
SMILES: OC1=CC=CN=C1C([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)