missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$44.28 - $44.28
Specifications
| CAS | 87199-18-6 |
|---|---|
| Melting Point | 213°C |
| Molecular Formula | C6H7BO3 |
| MDL Number | MFCD01074603 |
| InChI Key | WFWQWTPAPNEOFE-UHFFFAOYSA-N |
Specifications
| 87199-18-6 | |
| C6H7BO3 | |
| WFWQWTPAPNEOFE-UHFFFAOYSA-N | |
| 2734359 | |
| Crystalline Powder |
| 213°C | |
| MFCD01074603 | |
| (3-hydroxyphenyl)boronic acid | |
| 137.93 |