missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Hydroxymethyl)biphenyl 96.0+%, TCI Americaâ„¢
$264.57 - $791.81
Chemical Identifiers
CAS | 69605-90-9 |
---|---|
Molecular Formula | C13H12O |
Molecular Weight (g/mol) | 184.24 |
MDL Number | MFCD08703631 |
InChI Key | WGUZZTGZDPJWSG-UHFFFAOYSA-N |
Synonym | Biphenyl-3-methanol, 3-Phenylbenzyl Alcohol |
PubChem CID | 603556 |
IUPAC Name | {[1,1'-biphenyl]-3-yl}methanol |
SMILES | OCC1=CC=CC(=C1)C1=CC=CC=C1 |
Chemical Identifiers
69605-90-9 | |
184.24 | |
WGUZZTGZDPJWSG-UHFFFAOYSA-N | |
603556 | |
OCC1=CC=CC(=C1)C1=CC=CC=C1 |
C13H12O | |
MFCD08703631 | |
Biphenyl-3-methanol, 3-Phenylbenzyl Alcohol | |
{[1,1'-biphenyl]-3-yl}methanol |
Specifications
69605-90-9 | |
White-Yellow | |
MFCD08703631 | |
Biphenyl-3-methanol, 3-Phenylbenzyl Alcohol | |
OCC1=CC=CC(=C1)C1=CC=CC=C1 | |
184.24 | |
184.24 | |
Crystalline Powder |
50°C | |
C13H12O | |
1 g | |
WGUZZTGZDPJWSG-UHFFFAOYSA-N | |
{[1,1'-biphenyl]-3-yl}methanol | |
603556 | |
≥96.0% (GC) | |
3-(Hydroxymethyl)biphenyl |
Safety and Handling
TSCA : No