accessibility menu, dialog, popup

UserName

HO

CAS RN 69605-90-9

IUPAC Name: {[1,1'-biphenyl]-3-yl}methanol
Synonyms: [1,1'-biphenyl]-3-ylmethanol
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: WGUZZTGZDPJWSG-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results