missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)benzeneboronic acid pinacol ester

CAS: 443776-76-9 | C13H19BO3 | 234.102 g/mol

$68.14 - $850.88

Chemical Identifiers

CAS 443776-76-9
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.102
MDL Number MFCD09266196
InChI Key ZEWWJJQAFTXUIS-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
PubChem CID 44118236
IUPAC Name [3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5112603
View Documents
Thermo Scientific Chemicals
H5112603
1 g N/A
Only null left
 
AAH5112606
View Documents
Thermo Scientific Chemicals
H5112606
5 g N/A
Only null left
 
AAH5112614
View Documents
Thermo Scientific Chemicals
H5112614
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

443776-76-9
234.102
ZEWWJJQAFTXUIS-UHFFFAOYSA-N
44118236
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO
C13H19BO3
MFCD09266196
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

Specifications

443776-76-9
MFCD09266196
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO
234.102
234.1
C13H19BO3
1 g
ZEWWJJQAFTXUIS-UHFFFAOYSA-N
[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
44118236
3-(Hydroxymethyl)benzeneboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only