accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COH

CAS RN 443776-76-9

IUPAC Name: [3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
Synonyms: [3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
Molecular Weight (g/mol): 234.1
Molecular Formula: C13H19BO3
InChi Key: ZEWWJJQAFTXUIS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CO)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results