missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)benzeneboronic acid pinacol ester

Catalog No. AAH5112614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5112614 25 g
AAH5112603 1 g
AAH5112606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5112614 Supplier Thermo Scientific Chemicals Supplier No. H5112614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 443776-76-9 | C13H19BO3 | 234.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 443776-76-9
Molecular Formula C13H19BO3
Molecular Weight (g/mol) 234.102
MDL Number MFCD09266196
InChI Key ZEWWJJQAFTXUIS-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
PubChem CID 44118236
IUPAC Name [3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO

Specifications

Quantity 25 g
Formula Weight 234.1
Chemical Name or Material 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only