Learn More
3-Hydroxybenzyl alcohol, 97%
CAS: 620-24-6 | C7H8O2 | 124.139 g/mol
Supplier: Thermo Scientific Chemicals A1598114
Description
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1'-acetoxy-1-methyl-benzol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1′-acetoxy-1-methyl-benzol.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 3-Hydroxybenzyl alcohol | |
| 69°C to 72°C | |
| ∼300°C (decomposition) | |
| MFCD00004643 | |
| 2041500 | |
| Soluble in water. | |
| C1=CC(=CC(=C1)O)CO | |
| 124.139 | |
| CHEBI:17069 | |
| 98% |
| 620-24-6 | |
| 1.16 | |
| C7H8O2 | |
| 25 g | |
| 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol | |
| OKVJCVWFVRATSG-UHFFFAOYSA-N | |
| 3-(hydroxymethyl)phenol | |
| 102 | |
| 124.14 |
Chemical Identifiers
| 620-24-6 | |
| 124.139 | |
| OKVJCVWFVRATSG-UHFFFAOYSA-N | |
| 102 | |
| 3-(hydroxymethyl)phenol |
| C7H8O2 | |
| MFCD00004643 | |
| 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol | |
| CHEBI:17069 | |
| C1=CC(=CC(=C1)O)CO |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 210-633-6
RTECSNumber : DA4796700
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only