missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzyl alcohol, 97%

Catalog No. p-7022971
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598106 5 g
AAA1598114 25 g
AAA1598122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1598106 Supplier Thermo Scientific Chemicals Supplier No. A1598106
Only null left

CAS: 620-24-6 | C7H8O2 | 124.139 g/mol

3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1'-acetoxy-1-methyl-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1′-acetoxy-1-methyl-benzol.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 620-24-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004643
InChI Key OKVJCVWFVRATSG-UHFFFAOYSA-N
Synonym 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol
PubChem CID 102
ChEBI CHEBI:17069
IUPAC Name 3-(hydroxymethyl)phenol
SMILES C1=CC(=CC(=C1)O)CO

Specifications

Melting Point 69°C to 72°C
Density 1.16
Boiling Point ∼300°C (decomposition)
Quantity 5 g
Beilstein 2041500
Solubility Information Soluble in water.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material 3-Hydroxybenzyl alcohol
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 210-633-6
RTECSNumber DA4796700
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only