Learn More
3-Hydroxybenzonitrile, 99%
CAS: 873-62-1 | C7H5NO | 119.123 g/mol
Supplier: Thermo Scientific Chemicals B2170206
Description
3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzonitrile, is an intermediate that can be used for the synthesis of new Serotonin 5-HT1A receptor agonists having antinociceptive activity.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 3-Hydroxybenzonitrile | |
| 78°C to 81°C | |
| MFCD00002252 | |
| 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
| SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
| 3-hydroxybenzonitrile | |
| 13394 | |
| 99% |
| 873-62-1 | |
| C7H5NO | |
| 5 g | |
| Slightly soluble in water. | |
| C1=CC(=CC(=C1)O)C#N | |
| 119.123 | |
| 119.12 |
Chemical Identifiers
| 873-62-1 | |
| 119.123 | |
| SGHBRHKBCLLVCI-UHFFFAOYSA-N | |
| 13394 | |
| C1=CC(=CC(=C1)O)C#N |
| C7H5NO | |
| MFCD00002252 | |
| 3-cyanophenol, m-hydroxybenzonitrile, m-cyanophenol, benzonitrile, 3-hydroxy, benzonitrile, m-hydroxy, 3-hydroxy-benzonitrile, 3-cyano phenol, 3-cyano-phenol, 3-hydroxy benzonitrile, pubchem13113 | |
| 3-hydroxybenzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-845-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only