accessibility menu, dialog, popup

UserName

CAS RN 873-62-1

IUPAC Name: 3-hydroxybenzonitrile
Synonyms: M-cyanophenol
Molecular Weight (g/mol): 119.12
Molecular Formula: C7H5NO
InChi Key: SGHBRHKBCLLVCI-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)