missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyanthranilic acid, 97%

Catalog No. AC434870010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434870010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434870010 Supplier Thermo Scientific Chemicals Supplier No. 434870010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 548-93-6 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 548-93-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007700
InChI Key WJXSWCUQABXPFS-UHFFFAOYSA-N
Synonym 3-hydroxyanthranilic acid, 3-oxyanthranilic acid, 2-amino-3-hydroxy-benzoic acid, 3-hydroxy-anthranilsaeure, 3-ohaa, anthranilic acid, 3-hydroxy, 3-hydroxy anthranilic acid, benzoic acid, 2-amino-3-hydroxy, 3-hydroxyanthranilate, 3-hydroxy-anthranilsaeure german
PubChem CID 86
ChEBI CHEBI:15793
IUPAC Name 2-amino-3-hydroxybenzoic acid
SMILES NC1=C(O)C=CC=C1C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 224°C to 231°C
Color Colorless to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.14
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Hydroxyanthranilic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 208-962-5

RUO – Research Use Only