accessibility menu, dialog, popup

UserName

NH2OHHOO

CAS RN 548-93-6

IUPAC Name: 2-amino-3-hydroxybenzoic acid
Synonyms: 3-hydroxyanthranilic acid 3-hydroxy-anthranilsaeure 3-ohaa 3-hydroxyantranilic acid 3 hydroxyanthranilic acid acid, 3-hydroxyanthranilic 3-hydroxanthranilate 3-hydroxy-anthranilate 3-OH-anthranilic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: WJXSWCUQABXPFS-UHFFFAOYSA-N
SMILES: NC1=C(O)C=CC=C1C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)