Learn More
3'-Hydroxyacetophenone, 98%
CAS: 121-71-1 | C8H8O2 | 136.15 g/mol
$141.22 - $494.29
Chemical Identifiers
| CAS | 121-71-1 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00002298 |
| InChI Key | LUJMEECXHPYQOF-UHFFFAOYSA-N |
| Synonym | 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol |
| PubChem CID | 8487 |
| IUPAC Name | 1-(3-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC=C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1254722
|
Thermo Scientific Chemicals
A1254722 |
100 g |
Each for $141.22
|
|
|||||
|
AAA1254736
|
Thermo Scientific Chemicals
A1254736 |
500 g |
Each for $494.29
|
|
|||||
Description
3âHydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3âHydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and bases.
Chemical Identifiers
| 121-71-1 | |
| 136.15 | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 8487 | |
| CC(=O)C1=CC(=CC=C1)O |
| C8H8O2 | |
| MFCD00002298 | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| 1-(3-hydroxyphenyl)ethanone |
Specifications
| 121-71-1 | |
| 1.1 | |
| C8H8O2 | |
| 100 g | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 1-(3-hydroxyphenyl)ethanone | |
| 8487 | |
| 98% |
| 93°C to 97°C | |
| 296°C | |
| MFCD00002298 | |
| 2040676 | |
| Soluble in alcohol. Insoluble in water. | |
| CC(=O)C1=CC(=CC=C1)O | |
| 136.15 | |
| 136.15 | |
| 3'-Hydroxyacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-494-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only