missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetophenone, 98%

CAS: 121-71-1 | C8H8O2 | 136.15 g/mol

Supplier:  Thermo Scientific Chemicals A1254736

 View more versions of this product

Catalog No. AAA1254736


Only null left
Explore available promotions

Description

Description

3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

3'-Hydroxyacetophenone
93°C to 97°C
296°C
MFCD00002298
2040676
Soluble in alcohol. Insoluble in water.
CC(=O)C1=CC(=CC=C1)O
136.15
136.15
121-71-1
1.1
C8H8O2
500 g
3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
LUJMEECXHPYQOF-UHFFFAOYSA-N
1-(3-hydroxyphenyl)ethanone
8487
98%

Chemical Identifiers

121-71-1
136.15
LUJMEECXHPYQOF-UHFFFAOYSA-N
8487
CC(=O)C1=CC(=CC=C1)O
C8H8O2
MFCD00002298
3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
1-(3-hydroxyphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-494-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only