accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 121-71-1

IUPAC Name: 1-(3-hydroxyphenyl)ethan-1-one
Synonyms: 3-hydroxyacetophenone M-hydroxyacetophenone
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: LUJMEECXHPYQOF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(O)=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)