missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methylbenzoic acid, 98%

Catalog No. AAB2150114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2150114 25 g
AAB2150103 1 g
AAB2150106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2150114 Supplier Thermo Scientific Chemicals Supplier No. B2150114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 586-30-1 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 586-30-1
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002511
InChI Key ZQLCWPXBHUALQC-UHFFFAOYSA-N
Synonym 3-hydroxy-p-toluic acid, 3-hydroxy-4-methyl-benzoic acid, 3,4-cresotic acid, benzoic acid, 3-hydroxy-4-methyl, 3-hydroxy-4-methylbenzoicacid, pubchem3890, acmc-209m5x, 3-hydroxy-4-methyl benzoic acid, 3,4-cresotic acid; 3-hydroxy-p-toluic acid, 3-hydroxy-4-methylbenzoic acid, technical grade
PubChem CID 68512
IUPAC Name 3-hydroxy-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1O)C(O)=O

Specifications

Melting Point 207°C to 212°C
Odor Odorless
Quantity 25 g
Beilstein 1936497
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-571-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only