accessibility menu, dialog, popup

UserName

CH3HOOHO

CAS RN 586-30-1

IUPAC Name: 3-hydroxy-4-methylbenzoic acid
Synonyms: 3-hydroxy-4-methylbenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: ZQLCWPXBHUALQC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1O)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)